missing translation for 'onlineSavingsMsg'
Learn More

Sodium D-glucuronate monohydrate, 99%

Catalog No. AAL1398506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1398506 5 g
AAL1398514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1398506 Supplier Thermo Scientific Chemicals Supplier No. L1398506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 207300-70-7 ∣ C6H11NaO8 ∣ 234.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207300-70-7
Molecular Formula C6H11NaO8
Molecular Weight (g/mol) 234.14
MDL Number MFCD00151051
InChI Key ICQRTPLATMRWJI-UHFFFAOYNA-M
Synonym d-glucuronic acid sodium salt hydrate, glv8d2bupd, unii-glv8d2bupd, sodium d-glucuronate, sodium d-glucuronate monohydrate, sodium glucuronate hydrate jan, sodium 2s,3s,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate, sodium-d-glucuronate, d-glucuronic acid sodium salt
PubChem CID 24892836
SMILES O.[Na+].OC(C=O)C(O)C(O)C(O)C([O-])=O

Specifications

Melting Point 134°C to 138°C
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Beilstein 4218577
IUPAC Name sodium 2,3,4,5-tetrahydroxy-6-oxohexanoate hydrate
Formula Weight 234.14 (216.14 Anhydrous)
Percent Purity 99%
Grade Ungraded
Chemical Name or Material Sodium D-glucuronate monohydrate
EINECSNumber 239-065-7
RTECSNumber LZ8950000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only