missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanoborohydride, 95%

CAS: 25895-60-7 | CH3BNNa | 62.84 g/mol

Supplier:  Thermo Scientific Chemicals 168550500

 View more versions of this product

Catalog No. AC168550500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Sodium cyanoborohydride
25895-60-7
Powder
95%
NaBH3CN
04,448; 05,607; 06,537; 07,334; 08,454; 09,424; 10,360; 11,481; 12,445; 14,287; 16,305; 17,388
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
CVDUGUOQTVTBJH-UHFFFAOYSA-N
sodium cyanoboranuide
20587905
95%
>242.0°C
White
50 g
CH3BNNa
MFCD00003516
15, 8742
Solubility in water: 2120g/L at 29°C (slow decomposition). Other solubilities: soluble in methanol; soluble in thf, 372g/L, at 28°C, 410g/L at 46°C, 422g/L at 62°C, soluble in diglyme 176g/L at 25°C, slightly soluble in ethanol, insoluble in ether, b
[Na+].[BH3-]C#N
62.84
62.84
Glass bottle

Chemical Identifiers

25895-60-7
62.84
CVDUGUOQTVTBJH-UHFFFAOYSA-N
20587905
CH3BNNa
MFCD00003516
sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
[Na+].[BH3-]C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
In contact with water releases flammable gas.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic lif

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immedia

GHS Signal Word: Danger

EINECSNumber : 247-317-2

Recommended Storage : the substance is sensitive to gradually degrading on storage.

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only