missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanoborohydride, 95%

Catalog No. AC168550100
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC168550100 10 g Glass bottle
AC168550500 50 g Glass bottle
AC168552500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC168550100 Supplier Thermo Scientific Chemicals Supplier No. 168550100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25895-60-7 ∣ CH3BNNa ∣ 62.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent

Chemical Identifiers

CAS 25895-60-7
Molecular Formula CH3BNNa
Molecular Weight (g/mol) 62.84
MDL Number MFCD00003516
InChI Key CVDUGUOQTVTBJH-UHFFFAOYSA-N
Synonym sodium cyanoborohydride, sodium cyanotrihydridoborate, unii-c4i8c58p9t, zlchem 216, sodium cyanoboron 1-
PubChem CID 20587905
SMILES [Na+].[BH3-]C#N

Specifications

Melting Point >242.0°C
Color White
Physical Form Powder
Quantity 10 g
Packaging Glass bottle
Assay Percent Range 95%
Linear Formula NaBH3CN
Fieser 04,448; 05,607; 06,537; 07,334; 08,454; 09,424; 10,360; 11,481; 12,445; 14,287; 16,305; 17,388
Merck Index 15, 8742
Solubility Information Solubility in water: 2120g/L at 29°C (slow decomposition). Other solubilities: soluble in methanol; soluble in thf, 372g/L, at 28°C, 410g/L at 46°C, 422g/L at 62°C, soluble in diglyme 176g/L at 25°C, slightly soluble in ethanol, insoluble in ether, b
IUPAC Name sodium;cyanoboron(1-)
Formula Weight 62.84
Percent Purity 95%
Grade Ungraded
Chemical Name or Material Sodium cyanoborohydride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
In contact with water releases flammable gas.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Very toxic to aquatic lif
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immedia
EINECSNumber 247-317-2
Recommended Storage the substance is sensitive to gradually degrading on storage.

RUO – Research Use Only