missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Sodium Cyanoborohydride

Catalog No. 15615910G
Encompass
Change view
Click to view available options
Quantity:
10 g
50 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15615910G 10 g
1561591KG 1 kg
15615950G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15615910G Supplier Sigma Aldrich Supplier No. 15615910G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium Cyanoborohydride

Specifications

CAS 25895-60-7
Melting Point >242°C
Molecular Formula NaBH3CN
Linear Formula NaBH3CN
MDL Number MFCD00003516
Quantity 10 g
Synonym Sodium cyanotrihydridoborate
Molecular Weight (g/mol) 62.84 g/mol