missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, Reagent, ACS, 95%, Spectrum™ Chemical

Catalog No. 18606476
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606476 50 kg Poly Bottle
18606469 25 g Poly Bottle
18606470 125 g Poly Bottle
18606471 500 g Poly Bottle
18606473 2.5 kg Poly Bottle
18606472 6 x 500 g Poly Bottle
18606474 4 x 2.5 kg Poly Bottle
18606475 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18606476 Supplier Spectrum Chemical Mfg Cor Supplier No. S126050KGBL
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1260, 143-33-9, NaCN

Spectrum™ Chemical Sodium Cyanide, Reagent, ACS is a solid compound that is inorganic, water-soluble and white. A highly toxic salt, it is mainly used in gold mining because cyanide has a high affinity and reactivity with metals. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
IUPAC Name sodium iminomethanide
SMILES [Na+].[C-]#N

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Percent Purity 95%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Cyanide