missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, Reagent, ACS, 95%, Spectrum™ Chemical

S1260, 143-33-9, NaCN

Supplier:  Spectrum Chemical Mfg Cor S126012KG

 View more versions of this product

Catalog No. 18606475


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sodium Cyanide, Reagent, ACS is a solid compound that is inorganic, water-soluble and white. A highly toxic salt, it is mainly used in gold mining because cyanide has a high affinity and reactivity with metals. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Specifications

Specifications

143-33-9
12 kg
MFCD00003523
[Na+].[C-]#N
49.01
ACS
1
CNNa
MNWBNISUBARLIT-UHFFFAOYSA-N
sodium iminomethanide
95%
Fiber Drum

Chemical Identifiers

143-33-9
49.01
MNWBNISUBARLIT-UHFFFAOYSA-N
[Na+].[C-]#N
CNNa
MFCD00003523
sodium iminomethanide
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions