missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanide, granules, 98%, Thermo Scientific Chemicals

Catalog No. AAL1327818
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1327818 50 g
AAL1327830 250 g
AAL132780B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1327818 Supplier Thermo Scientific Chemicals Supplier No. L1327818
Only null left

Sodium cyanide is used as a starting material for the preparation of Reissert compounds and cyanogen bromide, cyanuric chloride and cyanogen chloride. It acts as a catalyst for the aminolysis of esters to primary amides. Furthermore, it is used for fumigation, electroplating, extracting gold and silver in mining industry. In organic synthesis, it is involved in the cyanation reaction of alkyl halides under phase transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium cyanide is used as a starting material for the preparation of Reissert compounds and cyanogen bromide, cyanuric chloride and cyanogen chloride. It acts as a catalyst for the aminolysis of esters to primary amides. Furthermore, it is used for fumigation, electroplating, extracting gold and silver in mining industry. In organic synthesis, it is involved in the cyanation reaction of alkyl halides under phase transfer conditions.

Solubility
Soluble in water, ammonia, methanol and ethanol. Slightly soluble in dimethylformamide. Insoluble in dimethylsulphoxide.

Notes
Hygroscopic. Do not store near acids. Incompatible with strong oxidizing agents and carbon dioxide.

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium;cyanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point ∼1500°C
Melting Point 563°C
Quantity 50 g
Linear Formula NaCN
UN Number UN1689
Beilstein 3587243
Merck Index 14,8605
Solubility Information Soluble in water,ammonia,methanol and ethanol. Slightly soluble in dimethylformamide. Insoluble in dimethylsulphoxide.
Formula Weight 49.01
Percent Purity 98%
Sensitivity Hygroscopic
Density 1.61
Chemical Name or Material Sodium cyanide, granules
Show More Show Less
Hazard Category H290-H300+H310+H330-H372
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P390-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SODIUM CYANIDE, SOLID
EINECSNumber 205-599-4
RTECSNumber VZ7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only