missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, ACS Reagent, Reagents

Catalog No. 01337736
Change view
Click to view available options
Quantity: 125 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01337736 Quantity: 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337736 Supplier Labchem Inc Supplier No. C1315500125D2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium Cyanide, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
IUPAC Name sodium iminomethanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point 1496°C
Melting Point 564°C
Color White
pH Alkaline
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.6
Chemical Name or Material Sodium Cyanide, ACS Reagent