missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Cyanide, ACS Reagent Grade, Ricca Chemical
$237.53 - $237.53
Chemical Identifiers
| CAS | 143-33-9 |
|---|---|
| Molecular Formula | CNNa |
| Molecular Weight (g/mol) | 49.01 |
| MDL Number | MFCD00003523 |
| InChI Key | MNWBNISUBARLIT-UHFFFAOYSA-N |
| IUPAC Name | sodium iminomethanide |
| SMILES | [Na+].[C-]#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
RDCS0470125
|
Ricca Chemical Company
RDCS0470125D2 |
125 g |
Each for $237.53
|
|
|||||
Chemical Identifiers
| 143-33-9 | |
| 49.01 | |
| MNWBNISUBARLIT-UHFFFAOYSA-N | |
| [Na+].[C-]#N |
| CNNa | |
| MFCD00003523 | |
| sodium iminomethanide |
Specifications
| 143-33-9 | |
| Fine Crystals or Crystalline Powder | |
| CNNa | |
| MNWBNISUBARLIT-UHFFFAOYSA-N | |
| sodium iminomethanide | |
| min 95% | |
| Amber Glass Bottle |
| 100 | |
| 125 g | |
| MFCD00003523 | |
| [Na+].[C-]#N | |
| 49.01 | |
| ACS | |
| Sodium Cyanide |
Safety and Handling
ShelfLife : 84 months