missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCS0470125
Change view
Click to view available options
Quantity:
125 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCS0470125 125 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCS0470125 Supplier Ricca Chemical Company Supplier No. RDCS0470125D2
Only null left

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
IUPAC Name sodium iminomethanide
SMILES [Na+].[C-]#N

Specifications

CAS Max % 100
Physical Form Fine Crystals or Crystalline Powder
Quantity 125 g
Percent Purity min 95%
Grade ACS
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Cyanide
ShelfLife 84 months