missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanide, ACS, 95% min, Thermo Scientific Chemicals

Catalog No. AA12137A3
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA12137A3 2 kg
AA1213722 100 g
AA1213736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA12137A3 Supplier Thermo Scientific Chemicals Supplier No. 012137A3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sodium cyanide is used as a starting material for the preparation of Reissert compounds, cyanogen bromide, cyanuric chloride and cyanogen chloride. It acts as a catalyst for the aminolysis of esters to primary amides. Furthermore, it is used for fumigation, in electroplating and for extracting gold and silver in mining industry. In organic synthesis, it is involved in the cyanation reaction of alkyl halides under phase transfer conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium;cyanide
SMILES [Na+].[C-]#N

Specifications

Boiling Point ca 1500°C
Melting Point 563°C
Color Gray
Physical Form Granules
Quantity 2 kg
Odor Weak, characteristic
Sensitivity Hygroscopic
Appearance White powder
Assay Percent Range 95% min.
Density 1.61
Beilstein 3587243
Solubility Information Soluble in water,ammonia,methanol and ethanol. Slightly soluble in dimethylformamide. Insoluble in dimethylsulphoxide.
Formula Weight 49.01
Percent Purity ≥95%
Grade ACS
Chemical Name or Material Sodium cyanide, For analysis ACS
Show More Show Less
Hazard Category H290-H300+H310+H330-H372
Precautionary Statement P234-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P312-P314-P330-P361-P363-P390-P501c
DOTInformation DOT Class: 6.1, Packaging Group: I
EINECSNumber 205-599-4
RTECSNumber VZ7525000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only