missing translation for 'onlineSavingsMsg'
Learn More

Sodium cyanide, 98%, for analysis, Thermo Scientific Chemicals

$176.58 - $176.58

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium iminomethanide
SMILES [Na+].[C-]#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC194665000
SDS View Documents
Thermo Scientific Chemicals
194665000
500 g Plastic bottle
Each for $176.58
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

143-33-9
49.01
MNWBNISUBARLIT-UHFFFAOYSA-N
8929
[Na+].[C-]#N
CNNa
MFCD00003523
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
sodium iminomethanide

Specifications

1497.0°C
143-33-9
Granular Crystalline Powder
97% min. (Argentometry)
NaCN
02,IV,5
15,8741
Solubility in water: 370g/L (20°C). Other solubilities: sparingly soluble in ethanol
[Na+].[C-]#N
49.01
49
Analytical
Sodium cyanide, p.a.
562.0°C
White
500 g
CNNa
MFCD00003523
04,446; 05,506; 06,535; 07,333; 08,453; 11,481; 13,185
sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
MNWBNISUBARLIT-UHFFFAOYSA-N
sodium iminomethanide
8929
98%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lastin

GHS P Statement
Absorb spillage to prevent material damage.
Wear protective gloves/protective clothing.
Rinse mouth.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a p

GHS Signal Word: Danger

EINECSNumber : 205-599-4

RTECSNumber : VZ7525000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only