missing translation for 'onlineSavingsMsg'
Learn More

Sodium Cyanide, 2.5% (w/v) (25 g/L), Ricca Chemical

Catalog No. 72414
Change view
Click to view available options
Quantity:
120 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
72414 120 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72414 Supplier Ricca Chemical Company Supplier No. 72414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 143-33-9
Molecular Formula CNNa
Molecular Weight (g/mol) 49.01
MDL Number MFCD00003523
InChI Key MNWBNISUBARLIT-UHFFFAOYSA-N
Synonym sodium cyanide, cyanogran, cymag, cyanide of sodium, sodium cyanide na cn, nacn, cyanobrik, cyanide salts, kyanid sodny, cyanasalt h
PubChem CID 8929
IUPAC Name sodium iminomethanide
SMILES [Na+].[C-]#N

Specifications

Concentration or Composition (by Analyte or Components) 2.5% (w/v) Aqueous Solution
Packaging Natural Poly Bottle
Quantity 120 mL
Chemical Name or Material Sodium Cyanide