missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Creatine Phosphate Hydrate 98.0+%, TCI America™
$46.60 - $402.63
Chemical Identifiers
| CAS | 922-32-7 |
|---|---|
| Molecular Formula | C4H8N3Na2O5P |
| Molecular Weight (g/mol) | 255.08 |
| MDL Number | MFCD00150192 |
| InChI Key | RNTXMYSPASRLFT-UHFFFAOYSA-L |
| Synonym | creatine phosphate sodium salt |
| PubChem CID | 131887655 |
| IUPAC Name | disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate |
| SMILES | [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O |
Chemical Identifiers
| 922-32-7 | |
| 255.08 | |
| RNTXMYSPASRLFT-UHFFFAOYSA-L | |
| 131887655 | |
| [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O |
| C4H8N3Na2O5P | |
| MFCD00150192 | |
| creatine phosphate sodium salt | |
| disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate |
Specifications
| White | |
| Crystalline Powder | |
| C4H8N3Na2O5P | |
| creatine phosphate sodium salt | |
| [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O | |
| 255.08 | |
| 255.08 | |
| Sodium Creatine Phosphate Hydrate |
| 922-32-7 | |
| 1 g | |
| MFCD00150192 | |
| RNTXMYSPASRLFT-UHFFFAOYSA-L | |
| disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate | |
| 131887655 | |
| ≥98.0% (HPLC) |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator