missing translation for 'onlineSavingsMsg'
Learn More

Sodium Creatine Phosphate Hydrate 98.0+%, TCI America™

Catalog No. C03971G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C03971G 1 g
C039725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C03971G Supplier TCI America Supplier No. C03971G

Chemical Identifiers

CAS 922-32-7
Molecular Formula C4H8N3Na2O5P
Molecular Weight (g/mol) 255.08
MDL Number MFCD00150192
InChI Key RNTXMYSPASRLFT-UHFFFAOYSA-L
Synonym creatine phosphate sodium salt
PubChem CID 131887655
IUPAC Name disodium ({amino[(carboxymethyl)(methyl)amino]methylidene}amino)phosphonate
SMILES [Na+].[Na+].CN(CC(O)=O)C(N)=NP([O-])([O-])=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 g
Formula Weight 255.08
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Sodium Creatine Phosphate Hydrate
TSCA Yes
Recommended Storage Refrigerator