missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate USP/FCC Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 6132-04-3
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
MK0734212
View Documents
Avantor Macron
073406
2.5kg Glass N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

6132-04-3
294.10
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
CHEBI:32142
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
C6H9Na3O9
NLJMYIDDQXHKNR-UHFFFAOYSA-K
71474
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate

Specifications

6132-04-3
trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
294.10
CHEBI:32142
FCC/USP
C6H9Na3O9
NLJMYIDDQXHKNR-UHFFFAOYSA-K
trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
71474
294.1
Sodium Citrate, Dihydrate
Videos
SDS
Documents

Documents