missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Dihydrate, Powder, U.S.P. - F.C.C., J.T. Baker™

Catalog No. p-5390907
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
02004068 500 g Poly Bottle
02004067 2.5 kg Poly Bottle
2 options

Catalog No. 02004068

Supplier: Avantor 365001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Physical Form Powder
Quantity 500 g
Formula Weight 294.1
Grade FCC/USP
Packaging Poly Bottle
Specific Gravity 1L = 1.70kg
Chemical Name or Material Sodium Citrate, Dihydrate