missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Sodium Citrate, Dihydrate, Molecular biology grade, Calbiochem™,

Catalog No. 5674465KG
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Fiber Carton
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5674465KG 5 kg Fiber Carton
567446100GM 100 g Glass Bottle
5674461KG 1 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 5674465KG Supplier MilliporeSigma™ Supplier No. 5674465KG

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
ChEBI CHEBI:32142

Specifications

Color White
Quantity 5 kg
Packaging Fiber Carton
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
PubChem CID 71474
Percent Purity ≥99% (Titration)
Grade Molecular Biology