missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Sodium Citrate, Dihydrate, Molecular biology grade, Calbiochem™,
Specifications
Specifications
| Color | White |
| Quantity | 1 kg |
| Packaging | Glass Bottle |
| SMILES | O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
| IUPAC Name | trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate |
| PubChem CID | 117063816 |
| Percent Purity | ≥99% (Titration) |
| Grade | Molecular Biology |
| Chemical Name or Material | Sodium Citrate |