missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate Dihydrate (Granular/USP/FCC/EP/BP/JP), Fisher Chemical™

Catalog No. p-2450865
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S470212 2.5 kg Poly Bottle
S47012 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S470212 Supplier Thermo Fisher Scientific Supplier No. S470212
Only null left

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
MDL Number MFCD00150031
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Alkalinity Pass Test (FCC/USP)
Acidity Pass Test (BP/EP)
Decomposition Information >230°C
Melting Point 300°C
Color White
Density 1.67g/mL
pH 7.5 to 9.5
Loss on Drying 10.0 to 13.0% (JP)
Packaging Poly Bottle
Linear Formula NaO2CCH2C(OH)(CO2Na)CH2CO2Na·2H2O
Quantity 2.5 kg
Residual Solvents Meets Requirements (BP/EP/USP)
Formula Weight 294.1
Percent Purity 99.0 to 101.0% (JP)
Grade BP/EP/FCC/JP/USP
Physical Form Solid
Chemical Name or Material Sodium Citrate Dihydrate
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0