missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Dihydrate, Granular, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. 18606460
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606460 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18606460 Supplier Spectrum Chemical Mfg Cor Supplier No. SO165500GM
Get Up to $500 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

SO165, 6132-04-3, C6H5Na3O7·2H2O

Spectrum™ Chemical Sodium Citrate, Dihydrate, Granular, USP is used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1545801
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.1
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
SMILES O.O.[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 500 g
Percent Purity 99 to 100.5%
Grade USP