missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate, Dihydrate, Granular, U.S.P. - F.C.C., J.T. Baker™
Specifications
Specifications
| Physical Form | Granules |
| Quantity | 500 g |
| Packaging | Wide Mouth Amber Glass Bottle |
| Specific Gravity | 1L = 1.70kg |
| Formula Weight | 294.1 |
| Percent Purity | 99.0% to 100.5% |
| Grade | FCC/USP |
| Chemical Name or Material | Sodium Citrate, Dihydrate |