missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Dihydrate, Granular, Multi-Compendial, U.S.P., J.T. Baker™

Catalog No. p-5390904
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
Catalog No. Quantity Packaging
02004056 500 g Wide Mouth Amber Glass Bottle
02004055 2.5 kg Poly Bottle
2 options

Catalog No. 02004056

Supplier: Avantor 364701

Only null left

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Physical Form Granules
Quantity 500 g
Formula Weight 294.1
Grade Multi-Compendial/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.70kg
Chemical Name or Material Sodium Citrate, Dihydrate