missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Dihydrate, Crystal, ACS, 99%, Spectrum™ Chemical

Catalog No. 18613457
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613457 50 kg Poly Bottle
18613454 500 g Poly Bottle
18613455 2.5 kg Poly Bottle
18613456 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613457 Supplier Spectrum Chemical Mfg Cor Supplier No. S125050KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6132-04-3

Specifications

pH 7.0 to 9.0
CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 50 kg
Molecular Weight (g/mol) 294.10
Percent Purity 99%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Citrate, Dihydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.