missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Dihydrate, ACS Grade, ≥ 99.0%, LabChem™

Catalog No. LC236501
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC236501 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC236501 Supplier LabChem Supplier No. LC236501

Chemical Identifiers

CAS 4-3-6132
Molecular Formula C6H9Na3O9
Molecular Weight (g/mol) 294.10
InChI Key NLJMYIDDQXHKNR-UHFFFAOYSA-K
Synonym trisodium citrate dihydrate, sodium citrate dihydrate, sodium citrate tribasic dihydrate, natriumcitrat-dihydrat, sodium citrate hydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, trisodium salt, dihydrate, citronensaeure-tri-na-salz-dihydrat, citric acid trisodium salt dihydrate, citric acid, trisodium salt dihydrate, trisodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate
PubChem CID 71474
ChEBI CHEBI:32142
IUPAC Name trisodium;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Na+].[Na+].[Na+]

Specifications

Melting Point 150°C
Color White
CAS Max % 100
Physical Form Crystals
Quantity 500 g
Linear Formula Na3C6O7H5·2H2O
Solubility Information Soluble in water
Formula Weight 294.1
Percent Purity ≥99.0%
Grade ACS
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.7g/cm3
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Density 1.7g/cm3
Chemical Name or Material Sodium Citrate, Dihydrate
Show More Show Less
Hazard Statement GHS H Statement
Product is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature