missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate, Anhydrous, USP, 99-100.5%, Spectrum™ Chemical
S1986, 68-04-2, C6H5Na3O7
Supplier: Spectrum Chemical Mfg Cor S198612KG
Description
Spectrum™ Chemical Sodium Citrate, Anhydrous, USP is used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 68-04-2 | |
| Fiber Drum | |
| MFCD00012462 | |
| HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
| trisodium 2-hydroxypropane-1,2,3-tricarboxylate | |
| 99 to 100.5% |
| 1 | |
| C6H5Na3O7 | |
| 12 kg | |
| [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O | |
| 258.07 | |
| USP |
Chemical Identifiers
| 68-04-2 | |
| 258.07 | |
| HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
| [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
| C6H5Na3O7 | |
| MFCD00012462 | |
| trisodium 2-hydroxypropane-1,2,3-tricarboxylate |