missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Anhydrous, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. 18606450
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
12 kg
2.5 kg
45 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18606450 2.5 kg Poly Pail
18606449 500 g Poly Bottle
18606451 12 kg Fiber Drum
18606453 12 kg Poly Pail
18606452 45 kg Fiber Drum
5 options

Catalog No. 18606450

Supplier: Spectrum Chemical Mfg Cor S19862.5KG

Only null left

S1986, 68-04-2, C6H5Na3O7

Spectrum™ Chemical Sodium Citrate, Anhydrous, USP is used to treat certain metabolic problems (acidosis) caused by kidney disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 68-04-2
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00012462
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg
Percent Purity 99 to 100.5%
Grade USP