missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Anhydrous, Powder, FCC, 99-100.5%, Spectrum™ Chemical

Catalog No. p-7158204
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606444 100 g Poly Bottle
18606445 500 g Poly Bottle
18606446 2.5 kg Poly Pail
18606447 12 kg Poly Pail
18606448 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18606444 Supplier Spectrum Chemical Mfg Cor Supplier No. S1254100GM
Only null left

S1254, 68-04-2, C6H5Na3O7

Spectrum™ Chemical Sodium Citrate, Anhydrous, Powder, FCC is used as a flavorant and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 68-04-2
Molecular Formula C6H5Na3O7
Molecular Weight (g/mol) 258.07
MDL Number MFCD00012462
InChI Key HRXKRNGNAMMEHJ-UHFFFAOYSA-K
IUPAC Name trisodium 2-hydroxypropane-1,2,3-tricarboxylate
SMILES [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

CAS Max % 1
Packaging Poly Bottle
Quantity 100 g
Percent Purity 99 to 100.5%
Grade FCC