missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate, Anhydrous, Powder, FCC, 99-100.5%, Spectrum™ Chemical
S1254, 68-04-2, C6H5Na3O7
Supplier: Spectrum Chemical Mfg Cor S1254500GM
Description
Spectrum™ Chemical Sodium Citrate, Anhydrous, Powder, FCC is used as a flavorant and preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 68-04-2 | |
| Poly Bottle | |
| MFCD00012462 | |
| HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
| trisodium 2-hydroxypropane-1,2,3-tricarboxylate | |
| 99 to 100.5% |
| 1 | |
| C6H5Na3O7 | |
| 500 g | |
| [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O | |
| 258.07 | |
| FCC |
Chemical Identifiers
| 68-04-2 | |
| 258.07 | |
| HRXKRNGNAMMEHJ-UHFFFAOYSA-K | |
| [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O |
| C6H5Na3O7 | |
| MFCD00012462 | |
| trisodium 2-hydroxypropane-1,2,3-tricarboxylate |