missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Citrate, Anhydrous, 99.0-100.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Quantity | 100 g |
| Percent Purity | 99 to 100.5% |
| Grade | Reagent |
| Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.