missing translation for 'onlineSavingsMsg'
Learn More

Sodium Citrate, Anhydrous, 99.0-100.5%, Spectrum™ Chemical

Catalog No. 18613453
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613453 2.5 kg Poly Bottle
18613449 100 g Poly Bottle
18613452 500 g Poly Bottle
18613450 12 kg Poly Bottle
18613451 45 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18613453 Supplier Spectrum Chemical Mfg Cor Supplier No. S12532.5KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 68-04-2

Specifications

CAS Max % 1
Physical Form Crystalline Powder or Needles
Quantity 2.5 kg
Molecular Weight (g/mol) 258.07
Percent Purity 99 to 100.5%
Grade Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Citrate, Anhydrous

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.