missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chromate Tetrahydrate, Crystal, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1245500GM

 View more versions of this product

Catalog No. 18613440


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

10034-82-9
500 g
Reagent
1
99%
Poly Bottle

Chemical Identifiers

10034-82-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.