missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chromate, Anhydrous, Reagents

Catalog No. p-200021109
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337034 100 g
01337033 3 kg
01337032 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337034 Supplier Labchem Inc Supplier No. C2314000100C2
Only null left

Sodium Chromate, Anhydrous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7775-11-3
Molecular Formula CrNa2O4
Molecular Weight (g/mol) 161.97
InChI Key PXLIDIMHPNPGMH-UHFFFAOYSA-N
Synonym Chromium disodium oxide
IUPAC Name disodium dioxochromiumbis(olate)
SMILES [Na+].[Na+].[O-][Cr]([O-])(=O)=O

Specifications

Melting Point 792°C
Color Yellow
Physical Form Solid
Quantity 100 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 2.723
Chemical Name or Material Sodium Chromate, Anhydrous