missing translation for 'onlineSavingsMsg'
Learn More

Sodium Chromate, Anhydrous, Reagents

Catalog No. 01337032
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
3 kg
500 g
Catalog No. Quantity
01337032 500 g
01337034 100 g
01337033 3 kg
3 options

Catalog No. 01337032

Supplier: Labchem Inc C2314000500A5

Only null left

Sodium Chromate, Anhydrous, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7775-11-3
Molecular Formula CrNa2O4
Molecular Weight (g/mol) 161.97
InChI Key PXLIDIMHPNPGMH-UHFFFAOYSA-N
Synonym Chromium disodium oxide
IUPAC Name disodium dioxochromiumbis(olate)
SMILES [Na+].[Na+].[O-][Cr]([O-])(=O)=O

Specifications

Melting Point 792°C
Color Yellow
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.723
Chemical Name or Material Sodium Chromate, Anhydrous