missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Chloride Standard, 1000 ppm NaCl, Ricca Chemical
$336.50 - $428.17
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-213-121
|
Ricca Chemical Company
R7223100500A |
500 mL |
Each for $336.50
|
|
|||||
|
R72231001A
|
Ricca Chemical Company
R72231001A |
1 L |
Each for $428.17
|
|
|||||
Chemical Identifiers
| 58.44 g/mol |
Specifications
| 100°C | |
| Colorless | |
| 99.9%,0.1% | |
| 500 mL | |
| 58.44 g/mol | |
| 1000 ppm | |
| Colorless Liquid |
| 0.0°C | |
| 7732-18-5,7647-14-5,7647-14-5,7732-18-5 | |
| Liquid | |
| 0.1%,99.9% | |
| Laboratory | |
| 500 mL HDPE Bottle | |
| Sodium Chloride Standard |
Safety and Handling
ShelfLife : 24 Months