missing translation for 'onlineSavingsMsg'
Learn More

Sodium chloride, pure, ca. 26% solution in water

CAS: 7647-14-5 | ClNa | 58.44 g/mol

$87.09 - $87.09

Specifications

Chemical Name or Material Sodium chloride
CAS 7647-14-5,7732-18-5
Color Colorless
Physical Form Liquid
Molecular Formula ClNa
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC387640010
View Documents
Thermo Scientific Chemicals
387640010
1 L Glass bottle
Each for $87.09
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium chloride
Colorless
ClNa
MFCD00003477
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
[Na+].[Cl-]
5234
58.44
25 to 28%
1.202
7647-14-5,7732-18-5
Liquid
NaCl
15,8734
FAPWRFPIFSIZLT-UHFFFAOYSA-M
58.44
CHEBI:26710
Pure
Glass bottle
1.2020g/mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only