missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sodium chloride, 5M aq. soln., RNAse free

Catalog No. AAJ60434AE
Change view
Click to view available options
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AAJ60434AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ60434AE Supplier Thermo Scientific™ Supplier No. J60434AE
Only null left

Specifications

CAS 7647-14-5
Physical Form Liquid
Quantity 100 mL
Molecular Formula ClNa
Linear Formula NaCl
MDL Number MFCD00003477
Formula Weight 58.44