missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99% min

Catalog No. p-4407871
Change view
Click to view available options
Quantity:
100 g
1 kg
5 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1426722 100 g
AA14267A1 1 kg
AA14267D9 5 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1426722 Supplier Thermo Scientific Chemicals Supplier No. 01426722

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7775-09-9 | ClNaO3 | 106.44 g/mol

Sodium chlorate is used to prepare chlorine dioxide in the pulp and paper industry. It is utilized for bleaching the wood pulp in order to make environmentally friendly and quality white paper products. It is also used to produce other perchlorate compounds in the industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
MDL Number MFCD00003479
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
IUPAC Name sodium chlorate
SMILES [Na+].[O-][Cl](=O)=O

Specifications

Melting Point 248°C
Color Colorless
Physical Form Granules
Quantity 100 g
Assay Percent Range 99% min.
Solubility Information Soluble in water,glycerol,hydrazine,acetone and methanol. Slightly soluble in ethanol and ammonia.
Formula Weight 106.45
Odor Odorless
Appearance Colorless granules
Density 2.5 g/cm3
Chemical Name or Material Sodium chlorate
Show More Show Less
Hazard Category H271-H302-H373
Precautionary Statement P210-P220-P221-P260-P264b-P270-P280-P283-P301+P312-P306+P360-P314-P330-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1, Packing Group: II
EINECSNumber 231-887-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only