missing translation for 'onlineSavingsMsg'
Learn More

Sodium chlorate, 99%, ACS reagent

Catalog No. AC446411000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446411000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC446411000 Supplier Thermo Scientific Chemicals Supplier No. 446411000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7775-09-9 ∣ ClNaO3 ∣ 106.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7775-09-9
Molecular Formula ClNaO3
Molecular Weight (g/mol) 106.44
InChI Key YZHUMGUJCQRKBT-UHFFFAOYSA-M
Synonym sodium chlorate, chloric acid, sodium salt, asex, chlorate de sodium, chlorsaure, tumbleleaf, atlacide, desolet, kusatol, rasikal
PubChem CID 516902
ChEBI CHEBI:65242
SMILES [Na+].[O-][Cl](=O)=O

Specifications

Melting Point 248.0°C to 261.0°C
Physical Form Crystalline Powder and/or Crystals
Quantity 100 g
Assay Percent Range 99%
Linear Formula NaClO3
Merck Index 15,8733
Solubility Information Solubility in water: 1000g/L (20°C). Other solubilities: soluble in alcohol and glycerin, 7.7g/L ethanol (20°C)
IUPAC Name sodium chlorate
Formula Weight 106.44
Percent Purity 99%
Grade ACS/Reagent
Packaging Plastic bottle
Specific Gravity 2.49
Density 2.4900g/mL
Chemical Name or Material Sodium chlorate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
May cause fire or explosion; strong oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
EINECSNumber 231-887-4

RUO – Research Use Only