missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate solution, Honeywell Fluka™
Volumetric, 0.5Â M Na2CO3 (1N)
$407.24 - $407.24
Chemical Identifiers
| CAS | 497-19-8 |
|---|---|
| Molecular Formula | CNa2O3 |
| Molecular Weight (g/mol) | 105.99 |
| MDL Number | MFCD00003494 |
| InChI Key | CDBYLPFSWZWCQE-UHFFFAOYSA-L |
| Synonym | sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda |
| PubChem CID | 10340 |
| ChEBI | CHEBI:29377 |
| SMILES | [Na+].[Na+].[O-]C([O-])=O |
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Chemical Identifiers
| 497-19-8 | |
| 105.99 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| 10340 | |
| [Na+].[Na+].[O-]C([O-])=O |
| CNa2O3 | |
| MFCD00003494 | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| CHEBI:29377 |
Specifications
| 497-19-8 | |
| CNa2O3 | |
| MFCD00003494 | |
| 4154566 | |
| CDBYLPFSWZWCQE-UHFFFAOYSA-L | |
| disodium;carbonate | |
| 10340 | |
| 105.99g/mol |
| 1 L | |
| Na2CO3 | |
| NONH for all modes of transport | |
| sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda | |
| [Na+].[Na+].[O-]C([O-])=O | |
| 105.99 | |
| CHEBI:29377 | |
| Sodium carbonate solution |