missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, Puriss., meets analytical spec. of BP, Ph. Eur., NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance), Honeywell Fluka™

Catalog No. p-7071805
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001981 1 kg
6001983 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001981 Supplier Honeywell Chemicals Supplier No. 134181KG
Only null left

Puriss., meets analytical specification of BP, Ph. Eur., NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 497-19-8
Molecular Formula CNa2O3
Molecular Weight (g/mol) 105.99
MDL Number MFCD00003494
InChI Key CDBYLPFSWZWCQE-UHFFFAOYSA-L
Synonym sodium carbonate, disodium carbonate, soda ash, carbonic acid disodium salt, calcined soda, sodium carbonate, anhydrous, carbonic acid, disodium salt, washing soda, sodiumcarbonate, solvay soda
PubChem CID 10340
ChEBI CHEBI:29377
SMILES [Na+].[Na+].[O-]C([O-])=O

Specifications

Physical Form Anhydrous
Quantity 1 kg
Linear Formula Na2CO3
UN Number NONH for all modes of transport
Beilstein 4154566
IUPAC Name disodium;carbonate
Formula Weight 105.99g/mol
Percent Purity 99.5 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Sodium Carbonate, Meets analytical specification of Ph.Eur., BP, NF, FCC
Hazard Statement P305 + P351 + P338
Precautionary Statement H319
EINECSNumber 207-838-8
RTECSNumber VZ4050000