missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Monohydrate, Granular, ACS, 99.5%, Spectrum™ Chemical

Catalog No. 18613376
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613376 500 g Poly Bottle
18613377 2.5 kg Poly Bottle
18613378 12 kg Poly Pail
18613379 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613376 Supplier Spectrum Chemical Mfg Cor Supplier No. S1230500GM
Only null left

Chemical Identifiers

CAS 5968-11-6

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99.5%
Grade ACS
Loss on Drying 13.0-15.0%
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.