missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Monohydrate, FCC, 99.5-100.5%, Spectrum™ Chemical

S1212, 5968-11-6, Na2CO3·H2O

$552.71 - $3837.64

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CH2Na2O4
Molecular Weight (g/mol) 124.00
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
IUPAC Name disodium hydrate carbonate
SMILES O.[Na+].[Na+].[O-]C([O-])=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18606390
View Documents
Spectrum Chemical Mfg Cor
S1212500GM
500 g Poly Bottle
Each for $552.71
Only null left
 
18606391
View Documents
Spectrum Chemical Mfg Cor
S121225KG
2.5 kg Poly Pail
Each for $1,620.57
Only null left
 
18606392
View Documents
Spectrum Chemical Mfg Cor
S121212KG
12 kg Poly Pail
Each for $3,837.64
Only null left
 
Description

Description

Spectrum™ Chemical Sodium Carbonate, Monohydrate, FCC is a food additive (E500) used as an acidity regulator, anti-caking agent, raising agent, and stabilizer. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

5968-11-6
124.00
disodium hydrate carbonate
CH2Na2O4
MQRJBSHKWOFOGF-UHFFFAOYSA-L
O.[Na+].[Na+].[O-]C([O-])=O

Specifications

5968-11-6
500 g
MQRJBSHKWOFOGF-UHFFFAOYSA-L
disodium hydrate carbonate
99.5 to 100.5%
12-15%
1
CH2Na2O4
O.[Na+].[Na+].[O-]C([O-])=O
124.00
FCC
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'