Learn More
Sodium carbonate monohydrate, ACS, 99.5% min
CAS: 5968-11-6 | CH2Na2O4 | 124.00 g/mol
Supplier: Thermo Scientific Chemicals 03648536
Description
It is widely employed as a water softener, an electrolyte, as a good conductor in electrolysis, as a primary standard for acid-base titrations and mostly as a buffer component in applications such as chromatography, capillary electrophoresis and enzyme catalysis. Also used in glass manufacture, detergent manufacture, sodium chemicals, carbonate chemicals manufacture, pulp and paper, brine treatment, flue gas desulfurization, coal treatment and ion exchange resin regeneration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Sodium carbonate monohydrate | |
| For analysis ACS | |
| Granules | |
| 99.5% | |
| MFCD00149177 | |
| Soluble in water and glycerol. Insoluble in alcohol,acetone,alkyl acetates,benzonitrile and liquid ammonia. | |
| C(=O)([O-])[O-].O.[Na+].[Na+] | |
| 124.00 | |
| 124.01 (105.99 Anhydrous) | |
| Hygroscopic |
| 100°C-H2O | |
| 5968-11-6 | |
| 500 g | |
| CH2Na2O4 | |
| sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
| MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
| disodium;carbonate;hydrate | |
| 2735133 | |
| ACS Reagent | |
| 2.250 g/mL |
Chemical Identifiers
| 5968-11-6 | |
| 124.00 | |
| MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
| 2735133 | |
| C(=O)([O-])[O-].O.[Na+].[Na+] |
| CH2Na2O4 | |
| MFCD00149177 | |
| sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
| disodium;carbonate;hydrate |
Safety and Handling
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H336
EINECSNumber : 207-838-8
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only