missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate monohydrate, ACS, 99.5% min

Catalog No. AA3648522
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3648522 100 g
AA3648536 500 g
AA36485A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3648522 Supplier Thermo Scientific Chemicals Supplier No. 03648522
Only null left

CAS: 5968-11-6 | CH2Na2O4 | 124.00 g/mol

It is widely employed as a water softener, an electrolyte, as a good conductor in electrolysis, as a primary standard for acid-base titrations and mostly as a buffer component in applications such as chromatography, capillary electrophoresis and enzyme catalysis. Also used in glass manufacture, detergent manufacture, sodium chemicals, carbonate chemicals manufacture, pulp and paper, brine treatment, flue gas desulfurization, coal treatment and ion exchange resin regeneration.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5968-11-6
Molecular Formula CH2Na2O4
Molecular Weight (g/mol) 124.00
MDL Number MFCD00149177
InChI Key MQRJBSHKWOFOGF-UHFFFAOYSA-L
Synonym sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop
PubChem CID 2735133
IUPAC Name disodium;carbonate;hydrate
SMILES C(=O)([O-])[O-].O.[Na+].[Na+]

Specifications

Melting Point 100°C-H2O
Physical Form Granules
Quantity 100 g
Assay Percent Range 99.5%
Solubility Information Soluble in water and glycerol. Insoluble in alcohol,acetone,alkyl acetates,benzonitrile and liquid ammonia.
Formula Weight 124.01 (105.99 Anhydrous)
Grade ACS Reagent
Sensitivity Hygroscopic
Density 2.250 g/mL
Chemical Name or Material Sodium carbonate monohydrate, For analysis ACS
Hazard Category H319-H336
Precautionary Statement P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
EINECSNumber 207-838-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only