missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate Monohydrate, ≥99.5%, ACS Reagent, Honeywell™ Fluka™
ACS Reagent,≥99.5%
$280.95 - $280.95
Chemical Identifiers
| CAS | 5968-11-6 |
|---|---|
| Molecular Formula | CH2Na2O4 |
| Molecular Weight (g/mol) | 124.00 |
| MDL Number | MFCD00149177 |
| InChI Key | MQRJBSHKWOFOGF-UHFFFAOYSA-L |
| Synonym | sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop |
| PubChem CID | 2735133 |
| IUPAC Name | disodium;carbonate;hydrate |
| SMILES | C(=O)([O-])[O-].O.[Na+].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 5968-11-6 | |
| 124.00 | |
| MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
| 2735133 | |
| C(=O)([O-])[O-].O.[Na+].[Na+] |
| CH2Na2O4 | |
| MFCD00149177 | |
| sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
| disodium;carbonate;hydrate |
Specifications
| 851°C | |
| 5968-11-6 | |
| CH2Na2O4 | |
| MFCD00149177 | |
| sodium carbonate monohydrate, disodium carbonate monohydrate, sodium carbonate, monohydrate, carbonic acid disodium salt, monohydrate, sodium carbonate hydrate, carbonic acid sodium salt monohydrate, disodium carbonate hydrate, disodium hydrate carbonate, na2co3 water, acmc-20ajop | |
| C(=O)([O-])[O-].O.[Na+].[Na+] | |
| 124.00 | |
| 124g/mol | |
| ACS Reagent | |
| Powder | |
| Sodium Carbonate Monohydrate |
| White | |
| 500 g | |
| Na2CO3 · H2O | |
| NONH for all modes of transport | |
| MQRJBSHKWOFOGF-UHFFFAOYSA-L | |
| disodium;carbonate;hydrate | |
| 2735133 | |
| ≥99.5% | |
| Poly Bottle | |
| 2.25g/cm3 |
Safety and Handling
P305 + P351 + P338
H319
EINECSNumber : 207-838-8