missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium carbonate decahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.), Honeywell Fluka™
Puriss., meets analytical specification of BP, FCC, Ph. Eur., E500, 99.5-100.5% (ex dried subst.)
$157.76 - $823.18
Chemical Identifiers
| CAS | 2-1-6132 |
|---|---|
| Molecular Formula | CH20Na2O13 |
| Molecular Weight (g/mol) | 286.14 |
| MDL Number | MFCD00149178 |
| InChI Key | XYQRXRFVKUPBQN-UHFFFAOYSA-L |
| Synonym | sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t |
| PubChem CID | 151402 |
| IUPAC Name | disodium;carbonate;decahydrate |
| SMILES | C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 2-1-6132 | |
| 286.14 | |
| XYQRXRFVKUPBQN-UHFFFAOYSA-L | |
| 151402 | |
| C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] |
| CH20Na2O13 | |
| MFCD00149178 | |
| sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t | |
| disodium;carbonate;decahydrate |
Specifications
| 2-1-6132 | |
| CH20Na2O13 | |
| MFCD00149178 | |
| sodium carbonate decahydrate, natrii carbonas decahydricus, carbonic acid disodium salt, decahydrate, unii-ls505bg22i, disodium decahydrate carbonate, acmc-1b9jt, natrium-carbonat-dekahydrat, natrium-carbonat-10-wasser, disodium carbonate decahydrate, ksc353a3t | |
| C(=O)([O-])[O-].O.O.O.O.O.O.O.O.O.O.[Na+].[Na+] | |
| 286.14 | |
| 286.14g/mol | |
| puriss. |
| 1 kg | |
| Na2CO3 · 10H2O | |
| NONH for all modes of transport | |
| XYQRXRFVKUPBQN-UHFFFAOYSA-L | |
| disodium;carbonate;decahydrate | |
| 151402 | |
| 99.5 to 100.5% (ex dried subst.) | |
| Sodium Carbonate Decahydrate, Meets analytical specification of Ph. Eur.BP, FCC |
Safety and Handling
P305 + P351 + P338
H319
EINECSNumber : 207-838-8