missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Carbonate, Decahydrate, 99.0-101.0%, Spectrum™ Chemical
Specifications
Specifications
| Melting Point | 32° to 38°C |
| CAS Max % | 1 |
| Quantity | 2.5 kg |
| Percent Purity | 99 to 101% |
| Grade | Reagent |
| Packaging | Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.