missing translation for 'onlineSavingsMsg'
Learn More

Sodium Carbonate, Decahydrate, 99.0-101.0%, Spectrum™ Chemical

Catalog No. 18613373
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 kg
50 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Catalog No. Quantity Packaging
18613373 500 g Poly Bottle
18613374 2.5 kg Poly Bottle
18613375 50 kg Fiber Drum
3 options

Catalog No. 18613373

Supplier: Spectrum Chemical Mfg Cor S1210500GM

Only null left

Chemical Identifiers

CAS 2-1-6132

Specifications

Melting Point 32° to 38°C
CAS Max % 1
Quantity 500 g
Percent Purity 99 to 101%
Grade Reagent
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.