missing translation for 'onlineSavingsMsg'
Learn More

Sodium carbonate, Concentrate Na2CO3 72 mM in Water, IC eluent, Concentrate (20x) for Metrosep A Supp 7, MilliporeSigma™ Supelco™

Catalog No. 111014322
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014322 2.5 L
111014321 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014322 Supplier Merck Emd Millipore Supplier No. 727842.5L

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This certified eluent concentrate for ion chromatography is traceable by potentiometric titration to NIST Standard Reference Material. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Specifications

Chemical Name or Material Sodium Carbonate
CAS 497-19-8
Physical Form Liquid
Quantity 2.5 L
Linear Formula Na2 CO3
MDL Number MFCD00003494
Synonym Sodium carbonate solution
Molecular Weight (g/mol) 105.99
Formula Weight 105.99
Grade Not Available
Concentration or Composition (by Analyte or Components) 72 mM Na2 CO3 aq. soln.
Show More Show Less